siRNA Details
| Gene Symbol: Olr831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000899.1 | NP_001000899.1 | 1 | 250 | ||
| Gene Symbol: Olr832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000898.1 | NP_001000898.1 | 1 | 250 | ||

Olr831
Genomic Map