siRNA Details
| Gene Symbol: Spint5p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008881.1 | NP_001008881.1 | 2 | 239 | ||
| XM_006235609.1 | XP_006235671.1 | 2 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spint5p| Gene Symbol: Spint5p | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008881.1 | NP_001008881.1 | 2 | 239 | ||
| XM_006235609.1 | XP_006235671.1 | 2 | 245 | |||