This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC497934 SIRNA Assays ›
Gene Symbol:
LOC497934
Gene Name:
similar to hypothetical protein FLJ20014
Gene Aliases:
-
Chromosome Location:
Chr.10: 55642070 - 55643924 on Build Rnor_6.0
UniGene ID:
Rn.45766
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s176227

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC497934
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017474.1 NP_001017474.2 1 703
GenBank mRNA BC082026.1 - 1 703 7129979

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028005, AK009886, AK169875, AK151629.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-vmxjx:80/100.66.76.90:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0