This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC497938 SIRNA Assays ›
Gene Symbol:
LOC497938
Gene Name:
similar to RIKEN cDNA 4933402P03
Gene Aliases:
-
Chromosome Location:
Chr.10: 56424164 - 56426012 on Build Rnor_6.0
UniGene ID:
Rn.195918
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s176237

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC497938
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017475.1 NP_001017475.1 3 748
XM_008767853.1 XP_008766075.1 2 777
XM_008767854.1 XP_008766076.1 2 777
GenBank mRNA BC079298.1 - 3 748 7134515

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-2m6b5:80/100.66.73.235:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0