siRNA Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109060.1 | NP_001102530.1 | 7 | 2165 | ||
| XM_006248743.2 | XP_006248805.1 | 7 | 2442 | |||
| XM_006248749.2 | XP_006248811.1 | 7 | 3060 | |||
| XM_008768989.1 | XP_008767211.1 | 5 | 2453 | |||
| XM_008768990.1 | XP_008767212.1 | 5 | 2447 | |||
| XM_008768991.1 | XP_008767213.1 | 4 | 1831 | |||
| GenBank mRNA | KF112845.1 | - | 7 | 2071 | ||
| KF112846.1 | - | 7 | 2071 | |||

Stard13
Genomic Map