This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC498368 SIRNA Assays ›
Gene Symbol:
LOC498368
Gene Name:
similar to RIKEN cDNA 0610040J01
Gene Aliases:
-
Chromosome Location:
Chr.14: 46050752 - 46134770 on Build Rnor_6.0
UniGene ID:
Rn.29370
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s176824

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC498368
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017500.1 NP_001017500.1 2 246
GenBank mRNA BC079124.1 - 2 246 7126862

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029554, BC004797, AK050792, AK002869, AK159779.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline