This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRC25 SIRNA Assays ›
Gene Symbol:
Lrrc25
Gene Name:
leucine rich repeat containing 25
Gene Aliases:
RGD1564818
Chromosome Location:
Chr.16: 20556973 - 20563772 on Build Rnor_6.0
UniGene ID:
Rn.126809
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s177123

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252977.2 XP_006253039.1 3 276
XM_006252978.2 XP_006253040.1 2 161
XM_008771187.1 XP_008769409.1 2 130
XM_008771188.1 XP_008769410.1 2 606
XM_573882.5 XP_573882.1 3 262
GenBank mRNA FQ228377.1 - 3 244

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gfvrm:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline