siRNA Details
| Gene Symbol: Tbc1d13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575106.5 | XP_575106.2 | 10 | 1094 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d13| Gene Symbol: Tbc1d13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575106.5 | XP_575106.2 | 10 | 1094 | ||