siRNA Details
| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025043.1 | NP_001020214.1 | 2 | 1692 | ||
| XM_006234927.2 | XP_006234989.1 | 2 | 2205 | |||
| GenBank mRNA | BC082013.1 | - | 2 | 1692 | 7132065 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpripl1| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025043.1 | NP_001020214.1 | 2 | 1692 | ||
| XM_006234927.2 | XP_006234989.1 | 2 | 2205 | |||
| GenBank mRNA | BC082013.1 | - | 2 | 1692 | 7132065 | |