siRNA Details
| Gene Symbol: Abhd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024314.1 | NP_001019485.1 | 12 | 1304 | ||
| GenBank mRNA | BC078918.1 | - | 12 | 1304 | 7096446 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abhd12| Gene Symbol: Abhd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024314.1 | NP_001019485.1 | 12 | 1304 | ||
| GenBank mRNA | BC078918.1 | - | 12 | 1304 | 7096446 | |