siRNA Details
| Gene Symbol: Tuba8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024339.1 | NP_001019510.1 | 4 | 1097 | ||
| XM_008763254.1 | XP_008761476.1 | 4 | 1234 | |||
| GenBank mRNA | BC079185.1 | - | 4 | 1097 | 7131106 | |
| FQ217798.1 | - | 4 | 1153 | |||

Tuba8
Genomic Map