siRNA Details
| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109267.1 | NP_001102737.1 | 2 | 193 | ||
| XM_006238968.2 | XP_006239030.1 | 2 | 213 | |||
| XM_006238969.2 | XP_006239031.1 | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc17| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109267.1 | NP_001102737.1 | 2 | 193 | ||
| XM_006238968.2 | XP_006239030.1 | 2 | 213 | |||
| XM_006238969.2 | XP_006239031.1 | 2 | 207 | |||