siRNA Details
Gene Symbol: Srf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109302.1 | NP_001102772.1 | 1 | 764 | ||
XM_006244557.2 | XP_006244619.1 | 1 | 767 | |||
XM_006244558.2 | XP_006244620.1 | 1 | 768 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_020493, BC051950.
Gene Ontology Categories:
