siRNA Details
| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024366.1 | NP_001019537.1 | 4 | 450 | ||
| XM_008773430.1 | XP_008771652.1 | 4 | 971 | |||
| GenBank mRNA | BC079325.1 | - | 4 | 450 | 7135460 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl13a| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024366.1 | NP_001019537.1 | 4 | 450 | ||
| XM_008773430.1 | XP_008771652.1 | 4 | 971 | |||
| GenBank mRNA | BC079325.1 | - | 4 | 450 | 7135460 | |