siRNA Details
| Gene Symbol: RGD1559970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773314.1 | XP_008771536.1 | 1 | 710 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559970| Gene Symbol: RGD1559970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773314.1 | XP_008771536.1 | 1 | 710 | ||