This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DEFA11 SIRNA Assays ›
Gene Symbol:
Defa11
Gene Name:
defensin alpha 11
Gene Aliases:
-
Chromosome Location:
Chr.16: 75517421 - 75519605 on Build Rnor_6.0
UniGene ID:
Rn.215839
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s181625

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Defa11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033078.1 NP_001028250.1 2 183
XM_006253369.1 XP_006253431.1 3 281
GenBank mRNA AY623755.1 - 2 183

Back To Top

More Information


Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s)

defense response to bacterium

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline