This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GDF3 SIRNA Assays ›
Gene Symbol:
GDF3
Gene Name:
growth differentiation factor 3
Gene Aliases:
KFS3, MCOP7, MCOPCB6
Chromosome Location:
Chr.12: 7689785 - 7695764 on Build GRCh38
UniGene ID:
Hs.86232
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s18391

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GDF3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020634.1 NP_065685.1 2 650
GenBank mRNA AF110641.1 - 2 625
AF263538.1 - 2 650
AY358875.1 - 2 679
BC030959.1 - 2 662 4610141

Back To Top

More Information


Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
eye development
in utero embryonic development
response to dietary excess
endoderm development
mesoderm development
regulation of cell fate commitment
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of BMP signaling pathway
notochord development
somite rostral/caudal axis specification
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of epidermal cell differentiation
negative regulation of myoblast differentiation
cell development
formation of anatomical boundary
SMAD protein signal transduction
primitive streak formation
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-zgjpq:80/100.66.78.143:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0