siRNA Details
| Gene Symbol: LOC302022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127530.1 | NP_001121002.1 | 1281 | |||
| GenBank mRNA | BC162000.1 | - | 1281 | 9036094 | ||
| Gene Symbol: LOC100911848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765047.1 | XP_008763269.1 | 3 | 927 | ||

LOC302022
Genomic Map