This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM45AL SIRNA Assays ›
Gene Symbol:
Tmem45al Tmem45a
Gene Name:
transmembrane protein 45A-like
transmembrane protein 45A
Gene Aliases:
-
-
Chromosome Location:
Chr.11: 46009220 - 46105670 on Build Rnor_6.0
Chromosome Location:
Chr.11: 46714401 - 46779496 on Build Rnor_6.0
UniGene ID:
-
Rn.198247
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s185229

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem45al
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751037.3 XP_003751085.1 4 986
XM_006248218.2 XP_006248280.1 4 989
XM_008768662.1 XP_008766884.1 4 990
XM_008768663.1 XP_008766885.1 4 986
Gene Symbol: Tmem45a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191074.1 NP_001178003.1 4 894

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-twvb7:80/100.66.73.158:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline