siRNA Details
| Gene Symbol: Spata13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191686.1 | NP_001178615.1 | 2 | 1233 | ||
| XM_006252113.2 | XP_006252175.1 | 2 | 1306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata13| Gene Symbol: Spata13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191686.1 | NP_001178615.1 | 2 | 1233 | ||
| XM_006252113.2 | XP_006252175.1 | 2 | 1306 | |||