siRNA Details
| Gene Symbol: Myeov2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109044.1 | NP_001102514.1 | 3 | 369 | ||
| GenBank mRNA | BC169124.1 | - | 3 | 372 | 6890894 | |
| DV717666.1 | - | 3 | 369 | |||
| FQ212403.1 | - | 3 | 361 | |||
| FQ221993.1 | - | 3 | 360 | |||
| FQ223515.1 | - | 3 | 360 | |||

Myeov2
Genomic Map