siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749786.3 | XP_003749834.1 | 5 | 722 | ||
| XM_006236797.2 | XP_006236859.1 | 4 | 581 | |||
| XM_008763097.1 | XP_008761319.1 | 5 | 738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pradc1| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749786.3 | XP_003749834.1 | 5 | 722 | ||
| XM_006236797.2 | XP_006236859.1 | 4 | 581 | |||
| XM_008763097.1 | XP_008761319.1 | 5 | 738 | |||