This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CALML4 SIRNA Assays ›
Gene Symbol:
Calml4
Gene Name:
calmodulin-like 4
Gene Aliases:
-
Chromosome Location:
Chr.8: 67753279 - 67765359 on Build Rnor_6.0
UniGene ID:
Rn.228661
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s189628

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Calml4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127575.1 NP_001121047.1 4 434
XM_006243243.2 XP_006243305.1 3 384
GenBank mRNA BC158759.1 - 4 386 7099908

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001102468, NM_138304, BC023475, AY061807, BY097052, and others.

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline