This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LARP7 SIRNA Assays ›
Gene Symbol:
Larp7
Gene Name:
La ribonucleoprotein domain family, member 7
Gene Aliases:
-
Chromosome Location:
Chr.2: 231866888 - 231882002 on Build Rnor_6.0
UniGene ID:
Rn.79854
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s189741

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Larp7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044290.2 NP_001037755.2 5 562
GenBank mRNA BC083898.1 - 5 499 7133376
FM031800.1 - 5 562

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138593, AK015041, BC020127, BB470802, AK162147, and others.

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
RNA processing
negative regulation of chromatin binding
germ cell proliferation
positive regulation of G1/S transition of mitotic cell cycle
nucleotide binding
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0