siRNA Details
| Gene Symbol: LOC102548695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228508.2 | XP_006228570.1 | 7 | 1390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548695| Gene Symbol: LOC102548695 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228508.2 | XP_006228570.1 | 7 | 1390 | ||