This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF146 SIRNA Assays ›
Gene Symbol:
Znf146 LOC102555183
Gene Name:
zinc finger protein 146
zinc finger protein OZF-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 88614736 - 88615684 on Build Rnor_6.0
Chromosome Location:
Chr.10: 74767528 - 74770722 on Build Rnor_6.0
UniGene ID:
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s190407

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Znf146
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759296.1 XP_008757518.1 1 412
Gene Symbol: LOC102555183
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247118.2 XP_006247180.1 2 1229

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: BC127039, AJ224763.

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-94nls:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0