This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other XLR3A SIRNA Assays ›
Gene Symbol:
Xlr3a LOC100911047
Gene Name:
X-linked lymphocyte-regulated 3A
X-linked lymphocyte-regulated protein 3A-like
Gene Aliases:
-
-
Chromosome Location:
Chr.1: 147033272 - 147049664 on Build Rnor_6.0
Chromosome Location:
Chr.1: 147000994 - 147012434 on Build Rnor_6.0
UniGene ID:
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s191227

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Xlr3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229630.2 XP_006229692.1 9 1667
XM_008759622.1 XP_008757844.1 9 1760
Gene Symbol: LOC100911047
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229628.2 XP_006229690.1 9 1668
XM_008759621.1 XP_008757843.1 9 1761

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4zngx:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline