This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PLPPR4 SIRNA Assays ›
Gene Symbol:
PLPPR4
Gene Name:
phospholipid phosphatase related 4
Gene Aliases:
LPPR4, LPR4, PHP1, PRG-1, PRG1
Chromosome Location:
Chr.1: 99262956 - 99309584 on Build GRCh38
UniGene ID:
Hs.13245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s19159

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PLPPR4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166252.1 NP_001159724.1 2, 3 561
NM_014839.4 NP_055654.2 2, 3 561
XM_011542498.2 XP_011540800.1 2, 3 509
GenBank mRNA AB007924.3 - 2, 3 557
AF357013.1 - 2, 3 334
AF541281.1 - 2, 3 568
AK291369.1 - 2, 3 361
AK300361.1 - 2, 3 305
AL834390.1 - 1, 2 74
AY304518.1 - 2, 3 509
BC042963.1 - 2, 3 379

Back To Top

More Information


Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
signal transduction
axonogenesis
phospholipid dephosphorylation
inner ear development
phosphatidate phosphatase activity
lipid phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-d7bgt:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline