siRNA Details
| Gene Symbol: Zbtb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001069858.5 | XP_001069858.2 | 2 | 2694 | ||
| XM_006234021.2 | XP_006234083.1 | 2 | 1723 | |||
| XM_006234022.2 | XP_006234084.1 | 2 | 2501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb34| Gene Symbol: Zbtb34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001069858.5 | XP_001069858.2 | 2 | 2694 | ||
| XM_006234021.2 | XP_006234083.1 | 2 | 1723 | |||
| XM_006234022.2 | XP_006234084.1 | 2 | 2501 | |||