siRNA Details
| Gene Symbol: Rps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105730.1 | NP_001099200.1 | 3 | 154 | ||
| GenBank mRNA | FQ210333.1 | - | 3 | 146 | ||
| FQ211223.1 | - | 3 | 146 | |||
| FQ217403.1 | - | 3 | 151 | |||
| FQ218290.1 | - | 3 | 145 | |||
| FQ221642.1 | - | 3 | 146 | |||
| FQ221735.1 | - | 3 | 147 | |||
| FQ222008.1 | - | 3 | 146 | |||
| FQ222172.1 | - | 3 | 146 | |||
| FQ222320.1 | - | 3 | 146 | |||
| FQ222499.1 | - | 3 | 146 | |||
| FQ222594.1 | - | 3 | 149 | |||
| FQ222598.1 | - | 3 | 147 | |||
| FQ222664.1 | - | 3 | 146 | |||
| FQ223680.1 | - | 3 | 147 | |||
| FQ224357.1 | - | 3 | 146 | |||
| FQ228386.1 | - | 3 | 147 | |||
| FQ228826.1 | - | 3 | 145 | |||
| FQ228989.1 | - | 3 | 146 | |||
| FQ229601.1 | - | 3 | 146 | |||
| X59277.1 | - | 3 | 135 | |||
| Gene Symbol: LOC100359503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728428.3 | XP_002728474.1 | 1 | 151 | ||

Rps28
Genomic Map