siRNA Details
| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083338.1 | NP_001076807.1 | 5 | 416 | ||
| XM_006250247.2 | XP_006250309.1 | 6 | 895 | |||
| GenBank mRNA | AB274732.1 | - | 5 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm40l| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083338.1 | NP_001076807.1 | 5 | 416 | ||
| XM_006250247.2 | XP_006250309.1 | 6 | 895 | |||
| GenBank mRNA | AB274732.1 | - | 5 | 416 | ||