This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TCL6 SIRNA Assays ›
Gene Symbol:
TCL6
Gene Name:
T-cell leukemia/lymphoma 6 (non-protein coding)
Gene Aliases:
TNG1, TNG2
Chromosome Location:
Chr.14: 95651178 - 95673452 on Build GRCh38
UniGene ID:
Hs.510368
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s195144

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TCL6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028288.1 7 1763
GenBank mRNA AB035332.1 - 4 3542
AB035333.1 - 3 3406
AB035334.1 - 5 2574
AB035335.1 - 4 2119
AB035337.1 - 7 1765
AB035338.1 - 7 1960
AB035339.1 - 2 231
AF195821.1 - 3 365
BX161462.1 - 9 1804

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bbcf2:80/100.66.78.33:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline