This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IP6K2 SIRNA Assays ›
Gene Symbol:
IP6K2
Gene Name:
inositol hexakisphosphate kinase 2
Gene Aliases:
IHPK2, PIUS
Chromosome Location:
Chr.3: 48688003 - 48717278 on Build GRCh38
UniGene ID:
Hs.595983
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s195221

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IP6K2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005909.2 NP_001005909.1 2 358
NM_001005910.2 NP_001005910.1 2 361
NM_001005911.2 NP_001005911.1 2 358
NM_001146178.2 NP_001139650.1 2 361
NM_001146179.2 NP_001139651.1 2 358
NM_001190316.1 NP_001177245.1 3 574
NM_001190317.1 NP_001177246.1 3 497
NM_016291.3 NP_057375.2 2 361
NR_027437.2 2 361
NR_027438.2 2 358
XM_006713199.3 XP_006713262.1 3 465
XM_006713200.1 XP_006713263.1 3 435
XM_006713201.1 XP_006713264.1 3 512
XM_006713202.1 XP_006713265.1 3 514
XM_011533816.1 XP_011532118.1 3 495
XM_011533817.1 XP_011532119.1 2 298
XM_011533818.1 XP_011532120.1 2 301
XM_011533822.1 XP_011532124.1 2 298
XM_011533823.1 XP_011532125.1 2 301
XM_017006583.1 XP_016862072.1 2 357
XM_017006584.1 XP_016862073.1 2 332
XM_017006585.1 XP_016862074.1 3 447
XM_017006586.1 XP_016862075.1 3 488
XM_017006587.1 XP_016862076.1 3 488
XM_017006588.1 XP_016862077.1 3 514
XM_017006589.1 XP_016862078.1 3 463
XM_017006590.1 XP_016862079.1 3 1069
XM_017006591.1 XP_016862080.1 3 434
XM_017006592.1 XP_016862081.1 3 463
XM_017006593.1 XP_016862082.1 3 463
GenBank mRNA AF177145.1 - 1 255
AK024045.1 - 2 298
AK097016.1 - 8 1216
AK290526.1 - 3 512
AK291501.1 - 3 489
AK304714.1 - 3 435
AK315204.1 - 2 299
AL137514.1 - 2 299
AY007091.1 - 2 284
BC001864.1 - 2 283
BC004469.2 - 2 288
BC019694.1 - 2 270 4941646
BC065533.1 - 2 287 6162048
BP262706.1 - 2 298
BX537544.1 - 3 513
CA488567.1 - 2 295
CD248129.1 - 2 324
CF139656.1 - 2 292
CN362780.1 - 2 273
DA083858.1 - 2 294
DB039955.1 - 2 358

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphate ion transport
negative regulation of cell growth
positive regulation of apoptotic process
inositol phosphate metabolic process
phosphatidylinositol phosphorylation
type I interferon signaling pathway
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
protein binding
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline