siRNA Details
| Gene Symbol: TOR4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017723.2 | NP_060193.2 | 2 | 434 | ||
| GenBank mRNA | AK000252.1 | - | 2 | 438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOR4A| Gene Symbol: TOR4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017723.2 | NP_060193.2 | 2 | 434 | ||
| GenBank mRNA | AK000252.1 | - | 2 | 438 | ||