This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRIM36 SIRNA Assays ›
Gene Symbol:
TRIM36
Gene Name:
tripartite motif containing 36
Gene Aliases:
HAPRIN, RBCC728, RNF98
Chromosome Location:
Chr.5: 115124762 - 115180546 on Build GRCh38
UniGene ID:
Hs.519514
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s195282

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIM36
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017397.1 NP_001017397.1 1 556
NM_001017398.1 NP_001017398.1 1 556
NM_001300752.1 NP_001287681.1 1 556
NM_018700.3 NP_061170.2 1 556
XM_017009621.1 XP_016865110.1 1 517
XM_017009623.1 XP_016865112.1 1 517
GenBank mRNA AB196558.1 - 1 47
AI142389.1 - 1 313
AJ272269.1 - 1 173
AK125426.1 - 1 333
BC045164.1 - 1 358
BC046096.1 - 1 504
BC130334.1 - 1 225 40146365

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
acrosome reaction
regulation of cell cycle
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-8p9zv:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline