This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AMN SIRNA Assays ›
Gene Symbol:
AMN
Gene Name:
amnion associated transmembrane protein
Gene Aliases:
PRO1028, amnionless
Chromosome Location:
Chr.14: 102922430 - 102930842 on Build GRCh38
UniGene ID:
Hs.534494
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s195422

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AMN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030943.3 NP_112205.2 12 1533
XM_011537202.2 XP_011535504.1 12 1778
XM_011537203.2 XP_011535505.1 12 2034
GenBank mRNA AF328788.1 - 12 1522
AK055802.1 - 11 2246

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
multicellular organism development
excretion
cobalamin metabolic process
cobalamin transport
lipoprotein metabolic process
Golgi to plasma membrane protein transport
receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0