siRNA Details
| Gene Symbol: CLEC4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244856.1 | NP_001231785.1 | 9 | 895 | ||
| NM_198492.3 | NP_940894.1 | 9 | 931 | |||
| GenBank mRNA | AY358431.1 | - | 10 | 928 | ||
| AY423706.1 | - | 9 | 931 | |||
| BC039679.1 | - | 5 | 518 | |||
| BC093691.1 | - | 9 | 885 | 7939536 | ||
| BC093693.1 | - | 9 | 885 | 7939538 | ||
| BC143309.1 | - | 9 | 849 | 9051814 | ||

CLEC4G
Genomic Map