siRNA Details
| Gene Symbol: KRTAP5-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012416.1 | NP_001012416.1 | 1 | 451 | ||
| GenBank mRNA | AB126075.1 | - | 1 | 451 | ||
| BC130399.1 | - | 1 | 423 | 40146430 | ||
| BC133035.1 | - | 1 | 423 | 40147057 | ||

KRTAP5-6
Genomic Map