This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CTRB2 SIRNA Assays ›
Gene Symbol:
CTRB2
Gene Name:
chymotrypsinogen B2
Gene Aliases:
-
Chromosome Location:
Chr.16: 75204096 - 75207174 on Build GRCh38
UniGene ID:
Hs.610926
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s196535

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CTRB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025200.3 NP_001020371.3 7 795
GenBank mRNA AK131056.1 - 7 782
AK225933.1 - 7 782
AK291822.1 - 7 780
BC073145.1 - 7 771 5747536

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
cobalamin metabolic process
extracellular matrix disassembly
serine-type endopeptidase activity
serine-type peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-bwr29:80/100.66.78.44:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0