This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ALYREF SIRNA Assays ›
Gene Symbol:
ALYREF
Gene Name:
Aly/REF export factor
Gene Aliases:
ALY, ALY/REF, BEF, REF, THOC4
Chromosome Location:
Chr.17: 81887835 - 81891622 on Build GRCh38
UniGene ID:
Hs.534385
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s19855

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ALYREF
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005782.3 NP_005773.3 2, 3 404
GenBank mRNA AF047002.1 - 2, 3 305
AF086513.1 - 1, 2 59
BC052302.1 - 2, 3 404 6381528

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011568, NM_019484, BC137658, U89876, BC099405, and others.

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA recombination
mRNA splicing, via spliceosome
osteoblast differentiation
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
mRNA 3'-end processing
replication fork processing
positive regulation of DNA-templated transcription, elongation
viral mRNA export from host cell nucleus
nucleotide binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline