This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPCR1 SIRNA Assays ›
Gene Symbol:
DPCR1
Gene Name:
diffuse panbronchiolitis critical region 1
Gene Aliases:
PBLT
Chromosome Location:
Chr.6: 30941000 - 30954221 on Build GRCh38
UniGene ID:
Hs.631993
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s199382

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DPCR1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080870.3 NP_543146.2 2 1456

Back To Top

More Information


Gene Ontology Categories:

Function(s)

positive regulation of protein targeting to mitochondrion

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-5djdz:80/100.66.79.206:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline