siRNA Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167947.2 | NP_001161419.1 | 8 | 1028 | ||
| NM_005834.4 | NP_005825.1 | 7 | 878 | |||
| GenBank mRNA | AF034790.1 | - | 7 | 879 | ||
| AF077039.1 | - | 6 | 744 | |||
| AJ005895.1 | - | 6 | 731 | |||
| BC010142.2 | - | 6 | 732 | 4300787 | ||

TIMM17B
Genomic Map