siRNA Details
| Gene Symbol: Acot3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134246.3 | NP_599007.1 | 4 | 1701 | ||
| GenBank mRNA | AY563097.1 | - | 4 | 1704 | ||
| AY563098.1 | - | 4 | 961 | |||
| BC138741.1 | - | 3 | 1300 | 8861763 | ||
| BC138743.1 | - | 3 | 1301 | 8861765 | ||

Acot3
Genomic Map