This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HNRNPA3 SIRNA Assays ›
Gene Symbol:
Hnrnpa3 Gm6793
Gene Name:
heterogeneous nuclear ribonucleoprotein A3
heterogeneous nuclear ribonucleoprotein A3 pseudogene
Gene Aliases:
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3
EG627828
Chromosome Location:
Chr.2: 75659230 - 75669407 on Build GRCm38
Chromosome Location:
Chr.8: 112013433 - 112015036 on Build GRCm38
UniGene ID:
Mm.316306
Mm.459502
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s202473

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnrnpa3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053263.1 NP_444493.1 10 1109
NM_146130.3 NP_666242.2 10 1175
NM_198090.2 NP_932758.1 10 1175
GenBank mRNA AB201711.1 - 10 1155
AF463524.1 - 10 1087
AK133860.1 - 10 1174
BC023828.1 - 10 1128 5343619
BC023908.1 - 10 1144 5322033
BC024454.1 - 7 692
BC038364.1 - 10 1157 5362605
BC062198.1 - 10 983 2655879
BC064824.1 - 10 1140 6477672
BC085474.1 - 9 1050 30538435
BC092083.1 - 7 770
BC116250.1 - 10 1078 40093600
BC116251.1 - 10 1078 40093602
BC157919.1 - 10 1078 9007347
BC158038.1 - 10 1078 9007341
Gene Symbol: Gm6793
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC157904.1 - 1 1060 9088095
BC158074.1 - 1 1060 9007371
BC172011.1 - 1 1060 9054297

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_194247, BC112027, AF517524, AK293125, BC064494, and others.
This siRNA also targets the following transcripts in Rat: NM_001111294, NM_001111295, NM_198132, XM_008761913, XM_008761911, and others.

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
mRNA transport
transmembrane transport
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
RNA transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-ghgff:80/100.66.76.23:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0