This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IL17D SIRNA Assays ›
Gene Symbol:
Il17d
Gene Name:
interleukin 17D
Gene Aliases:
AI462269, IL-17D, Il27a
Chromosome Location:
Chr.14: 57524829 - 57543166 on Build GRCm38
UniGene ID:
Mm.390726
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s202701

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il17d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145837.3 NP_665836.2 3 880
GenBank mRNA AF458063.1 - 2 802
AK131917.1 - 3 929
BC099514.1 - 1 322

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770785.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0