siRNA Details
| Gene Symbol: Cisd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025902.3 | NP_080178.1 | 2 | 322 | ||
| GenBank mRNA | AK003486.1 | - | 2 | 293 | ||
| AK004257.1 | - | 2 | 293 | |||
| AK005184.1 | - | 2 | 281 | |||
| AK005301.1 | - | 2 | 297 | |||
| AM162549.1 | - | 2 | 814 | |||
| BC058279.1 | - | 2 | 322 | 6528467 | ||

Cisd2
Genomic Map