siRNA Details
| Gene Symbol: Shisa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174086.1 | NP_001167557.1 | 1 | 810 | ||
| NM_028277.2 | NP_082553.2 | 1 | 810 | |||
| GenBank mRNA | AK012380.1 | - | 1 | 811 | ||
| AK049860.1 | - | 1 | 336 | |||
| BC030884.1 | - | 1 | 99 | |||
| GU479981.1 | - | 1 | 342 | |||
| GU479982.1 | - | 1 | 342 | |||

Shisa9
Genomic Map