This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700037C18RIK SIRNA Assays ›
Gene Symbol:
1700037C18Rik
Gene Name:
RIKEN cDNA 1700037C18 gene
Gene Aliases:
2810022L20Rik
Chromosome Location:
Chr.16: 3905798 - 3908690 on Build GRCm38
UniGene ID:
Mm.486226
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s206506

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700037C18Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028484.2 NP_082760.2 3 713
GenBank mRNA AK012792.1 - 3 683
AK161085.1 - 720
BC147295.1 - 3 501 9056323

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001080524.
This siRNA also targets the following transcripts in Rat: XM_006245837, XM_340757, XM_006245836.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline