siRNA Details
Gene Symbol: 1700010D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005817.1 | - | 1, 2 | 290 | ||
BC059715.1 | - | 1, 2 | 293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700010D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005817.1 | - | 1, 2 | 290 | ||
BC059715.1 | - | 1, 2 | 293 |