This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FLYWCH2 SIRNA Assays ›
Gene Symbol:
Flywch2
Gene Name:
FLYWCH family member 2
Gene Aliases:
2810417J12Rik, 3010014L17Rik, AW548203
Chromosome Location:
Chr.17: 23776918 - 23786077 on Build GRCm38
UniGene ID:
Mm.269613
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s206778

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Flywch2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029798.3 NP_084074.1 2 332
GenBank mRNA AK013108.1 - 2 334
AK019378.1 - 2 320
BC024947.1 - 2 288 2648415

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008767601.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0